siRNA Details
| Gene Symbol: ASH1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027023.1 | 1 | 1626 | |||
| GenBank mRNA | AK126559.1 | - | 1 | 1577 | ||
| BC017347.2 | - | 2 | 571 | |||
| BC065229.1 | - | 1 | 347 | |||
| BC104640.1 | - | 1 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASH1L-AS1| Gene Symbol: ASH1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027023.1 | 1 | 1626 | |||
| GenBank mRNA | AK126559.1 | - | 1 | 1577 | ||
| BC017347.2 | - | 2 | 571 | |||
| BC065229.1 | - | 1 | 347 | |||
| BC104640.1 | - | 1 | 365 | |||