siRNA Details
Gene Symbol: NEXN-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103535.1 | 2 | 1535 | |||
GenBank mRNA | AK075118.1 | - | 3 | 1516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000421331.1 | - | 2 | 1535 | Chr. 1 - 77881348 - 77889539 [-] on Build GRCh38 |
