siRNA Details
| Gene Symbol: TTC28-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026963.1 | 5 | 2207 | |||
| GenBank mRNA | AB051435.1 | - | 4 | 1925 | ||
| AL832597.1 | - | 5 | 2163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC28-AS1| Gene Symbol: TTC28-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026963.1 | 5 | 2207 | |||
| GenBank mRNA | AB051435.1 | - | 4 | 1925 | ||
| AL832597.1 | - | 5 | 2163 | |||