siRNA Details
Gene Symbol: UBOX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038395.1 | 6 | 1355 | |||
GenBank mRNA | AK097516.1 | - | 6 | 1355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446537.5 | - | 6 | 1353 | Chr. 20 - 3106913 - 3150867 [+] on Build GRCh38 |
