siRNA Details
| Gene Symbol: LINC00445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132115.1 | 2 | 292 | |||
| NR_132116.1 | 2 | 280 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422430.1 | - | 2 | 280 | Chr. 13 - 35697524 - 35699001 [+] on Build GRCh38 | |

LINC00445
Genomic Map