siRNA Details
| Gene Symbol: LOC101928335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110395.1 | 4 | 1612 | |||
| GenBank mRNA | AK094711.1 | - | 4 | 1612 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430140.1 | - | 4 | 1612 | Chr. X - 107894597 - 107935980 [-] on Build GRCh38 | |

LOC101928335
Genomic Map