siRNA Details
| Gene Symbol: ELFN1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120508.1 | 2 | 778 | |||
| NR_120509.1 | 2 | 581 | ||||
| NR_120510.1 | 2 | 445 | ||||
| GenBank mRNA | HY180852.1 | - | 2 | 446 | ||
| JX046910.1 | - | 2 | 428 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453348.1 | - | 2 | 407 | Chr. 7 - 1738630 - 1742272 [-] on Build GRCh38 | |

ELFN1-AS1
Genomic Map