siRNA Details
| Gene Symbol: LOC102724058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110598.1 | 3 | 332 | |||
| GenBank mRNA | HY008183.1 | - | 3 | 309 | ||
| HY011842.1 | - | 3 | 332 | |||
| HY029566.1 | - | 3 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724058| Gene Symbol: LOC102724058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110598.1 | 3 | 332 | |||
| GenBank mRNA | HY008183.1 | - | 3 | 309 | ||
| HY011842.1 | - | 3 | 332 | |||
| HY029566.1 | - | 3 | 320 | |||