siRNA Details
| Gene Symbol: MIR7515HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110497.1 | 1 | 357 | |||
| GenBank mRNA | BX119852.1 | - | 1 | 282 | ||
| DB061664.1 | - | 1 | 357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR7515HG| Gene Symbol: MIR7515HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110497.1 | 1 | 357 | |||
| GenBank mRNA | BX119852.1 | - | 1 | 282 | ||
| DB061664.1 | - | 1 | 357 | |||