siRNA Details
Gene Symbol: LINC01376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135287.1 | 4 | 1845 | |||
GenBank mRNA | AK123475.1 | - | 4 | 1847 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000424895.1 | - | 4 | 1845 | Chr. 2 - 18986451 - 19026971 [-] on Build GRCh38 |
