siRNA Details
| Gene Symbol: GACAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX107472.1 | - | 3 | 439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GACAT3| Gene Symbol: GACAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX107472.1 | - | 3 | 439 | ||