siRNA Details
| Gene Symbol: KCCAT333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110014.1 | 4 | 539 | |||
| NR_110015.1 | 3 | 471 | ||||
| GenBank mRNA | CB134096.1 | - | 3 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCCAT333| Gene Symbol: KCCAT333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110014.1 | 4 | 539 | |||
| NR_110015.1 | 3 | 471 | ||||
| GenBank mRNA | CB134096.1 | - | 3 | 371 | ||