siRNA Details
Gene Symbol: LOC153910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027311.1 | 3 | 1207 | |||
NR_027312.1 | 3 | 1210 | ||||
GenBank mRNA | AK055933.1 | - | 3 | 1210 | ||
BC033340.1 | - | 1 | 529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000447311.1 | - | 3 | 1210 | Chr. 6 - 142526455 - 142637889 [-] on Build GRCh38 |
