siRNA Details
| Gene Symbol: LINC00854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047479.2 | 5 | 906 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX118168.1 | - | 3 | 528 | Chr. 17 - 43217105 - 43224523 [-] on Build GRCh38 | |

LINC00854
Genomic Map