siRNA Details
| Gene Symbol: LOC101927476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110386.1 | 3 | 857 | |||
| GenBank mRNA | AK123782.1 | - | 3 | 857 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452690.1 | - | 3 | 857 | Chr. X - 40262917 - 40287720 [+] on Build GRCh38 | |

LOC101927476
Genomic Map