siRNA Details
Gene Symbol: LOC101927476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110386.1 | 3 | 2765 | |||
GenBank mRNA | AK123782.1 | - | 3 | 2764 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000452690.1 | - | 3 | 2765 | Chr. X - 40262917 - 40287720 [+] on Build GRCh38 |
