siRNA Details
| Gene Symbol: LOC654841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102371.1 | 8 | 981 | |||
| GenBank mRNA | AK056332.1 | - | 8 | 1047 | ||
| BC035052.2 | - | 8 | 982 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC654841| Gene Symbol: LOC654841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102371.1 | 8 | 981 | |||
| GenBank mRNA | AK056332.1 | - | 8 | 1047 | ||
| BC035052.2 | - | 8 | 982 | |||