siRNA Details
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 318 | |||
GenBank mRNA | BC042616.1 | - | 3 | 320 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 318 | |||
GenBank mRNA | BC042616.1 | - | 3 | 320 |