siRNA Details
| Gene Symbol: SIX3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103785.1 | 2 | 854 | |||
| NR_103786.1 | 2 | 703 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419364.1 | - | 2 | 698 | Chr. 2 - 44940154 - 44941873 [-] on Build GRCh38 | |

SIX3-AS1
Genomic Map