siRNA Details
| Gene Symbol: LOC101928201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110390.1 | 1, 2 | 68 | |||
| GenBank mRNA | BC038195.1 | - | 1, 2 | 68 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420725.1 | - | 1, 2 | 68 | Chr. X - 4627200 - 4633572 [-] on Build GRCh38 | |

LOC101928201
Genomic Map