siRNA Details
Gene Symbol: LINC01424 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109928.1 | 2 | 965 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417820.1 | - | 2 | 629 | Chr. 21 - 44802577 - 44804717 [+] on Build GRCh38 |
