siRNA Details
| Gene Symbol: LINC01424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109928.1 | 2 | 796 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417820.1 | - | 2 | 460 | Chr. 21 - 44802577 - 44804717 [+] on Build GRCh38 | |

LINC01424
Genomic Map