siRNA Details
| Gene Symbol: LOC101927973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126012.1 | 4 | 573 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453420.5 | - | 4 | 573 | Chr. 21 - 28116094 - 28228667 [-] on Build GRCh38 | |

LOC101927973
Genomic Map