siRNA Details
| Gene Symbol: RNF217-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026876.1 | 7 | 2578 | |||
| GenBank mRNA | AL713649.2 | - | 7 | 2580 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439075.1 | - | 7 | 2590 | Chr. 6 - 124909093 - 124963039 [-] on Build GRCh38 | |

RNF217-AS1
Genomic Map