siRNA Details
| Gene Symbol: LOC100508046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110505.1 | 2 | 2387 | |||
| GenBank mRNA | AY338953.1 | - | 2 | 2385 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422014.1 | - | 2 | 2384 | Chr. 22 - 15699361 - 15703403 [-] on Build GRCh38 | |

LOC100508046
Genomic Map