siRNA Details
| Gene Symbol: LOC100505588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135129.1 | 2 | 614 | |||
| GenBank mRNA | AK131516.1 | - | 2 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505588| Gene Symbol: LOC100505588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135129.1 | 2 | 614 | |||
| GenBank mRNA | AK131516.1 | - | 2 | 565 | ||