siRNA Details
| Gene Symbol: LOC100505588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135129.1 | 2 | 1325 | |||
| GenBank mRNA | AK131516.1 | - | 2 | 1282 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453529.2 | - | 2 | 1325 | Chr. 9 - 124006277 - 124009396 [-] on Build GRCh38 | |

LOC100505588
Genomic Map