siRNA Details
| Gene Symbol: LINC01010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038216.1 | 3 | 677 | |||
| NR_038217.1 | 3 | 468 | ||||
| NR_038218.1 | 3 | 434 | ||||
| NR_038219.1 | 5 | 647 | ||||
| GenBank mRNA | AK055166.1 | - | 3 | 471 | ||
| BC041589.1 | - | 3 | 406 | |||
| DA244667.1 | - | 3 | 423 | |||
| DA419376.1 | - | 3 | 471 | |||

LINC01010
Genomic Map