siRNA Details
| Gene Symbol: LINC01010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038216.1 | 3 | 1549 | |||
| NR_038217.1 | 3 | 1340 | ||||
| NR_038218.1 | 3 | 1306 | ||||
| NR_038219.1 | 5 | 1519 | ||||
| GenBank mRNA | BC041589.1 | - | 3 | 1278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01010| Gene Symbol: LINC01010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038216.1 | 3 | 1549 | |||
| NR_038217.1 | 3 | 1340 | ||||
| NR_038218.1 | 3 | 1306 | ||||
| NR_038219.1 | 5 | 1519 | ||||
| GenBank mRNA | BC041589.1 | - | 3 | 1278 | ||