siRNA Details
| Gene Symbol: LOC101926975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110558.1 | 4 | 1030 | |||
| GenBank mRNA | AL833138.1 | - | 4 | 1031 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432677.1 | - | 4 | 1030 | Chr. 5 - 142745600 - 142760998 [+] on Build GRCh38 | |

LOC101926975
Genomic Map