siRNA Details
| Gene Symbol: OVAAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125716.1 | 3 | 1404 | |||
| GenBank mRNA | BC036830.1 | - | 3 | 1401 | ||
| BC044260.1 | - | 3 | 1402 | |||
| BC063624.1 | - | 3 | 1404 | |||
| BC073892.1 | - | 3 | 1428 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000442621.1 | - | 3 | 1402 | Chr. 1 - 180558976 - 180566518 [+] on Build GRCh38 | |

OVAAL
Genomic Map