siRNA Details
Gene Symbol: LOC101927189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125842.1 | 4 | 2777 | |||
GenBank mRNA | AK056584.1 | - | 4 | 2578 | ||
BX640790.1 | - | 4 | 2712 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431554.2 | - | 4 | 2713 | Chr. 6 - 53930046 - 53997667 [+] on Build GRCh38 |
