siRNA Details
| Gene Symbol: LOC101927189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125842.1 | 4 | 911 | |||
| GenBank mRNA | AK056584.1 | - | 4 | 712 | ||
| BX640790.1 | - | 4 | 850 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431554.2 | - | 4 | 847 | Chr. 6 - 53930046 - 53997667 [+] on Build GRCh38 | |

LOC101927189
Genomic Map