siRNA Details
Gene Symbol: LINC01186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110388.1 | 2 | 546 | |||
GenBank mRNA | AL700385.1 | - | 2 | 514 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000446884.1 | - | 2 | 517 | Chr. X - 46325924 - 46327645 [-] on Build GRCh38 |
