siRNA Details
| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038373.1 | 4 | 1265 | |||
| NR_134458.1 | 3 | 1445 | ||||
| GenBank mRNA | BC043233.1 | - | 4 | 1273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC9-AS1| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038373.1 | 4 | 1265 | |||
| NR_134458.1 | 3 | 1445 | ||||
| GenBank mRNA | BC043233.1 | - | 4 | 1273 | ||