siRNA Details
| Gene Symbol: LOC644838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038844.1 | 8 | 1132 | |||
| GenBank mRNA | AK123766.1 | - | 8 | 1132 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433133.5 | - | 8 | 1132 | Chr. 2 - 67123357 - 67215319 [-] on Build GRCh38 | |

LOC644838
Genomic Map