siRNA Details
Gene Symbol: LOC101928700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110829.1 | 4 | 1257 | |||
GenBank mRNA | BC026117.1 | - | 4 | 1262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434887.1 | - | 4 | 1257 | Chr. 7 - 147080934 - 147097609 [-] on Build GRCh38 |
