siRNA Details
| Gene Symbol: LINC00529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ301561.1 | - | 2 | 148 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443854.1 | - | 2 | 148 | Chr. 8 - 11247626 - 11267865 [-] on Build GRCh38 | |

LINC00529
Genomic Map