siRNA Details
| Gene Symbol: LOC284898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318329.1 | NP_001305258.1 | 2 | 567 | ||
| GenBank mRNA | BC036876.1 | - | 2 | 554 | ||
| DA498162.1 | - | 2 | 567 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453934.1 | - | 2 | 567 | Chr. 22 - 27048144 - 27060518 [-] on Build GRCh38 | |

LOC284898
Genomic Map