siRNA Details
| Gene Symbol: NIFK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037856.1 | 3 | 806 | |||
| NR_037858.1 | 2 | 1338 | ||||
| GenBank mRNA | BC036624.2 | - | 2 | 879 | ||
| BC047384.1 | - | 3 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIFK-AS1| Gene Symbol: NIFK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037856.1 | 3 | 806 | |||
| NR_037858.1 | 2 | 1338 | ||||
| GenBank mRNA | BC036624.2 | - | 2 | 879 | ||
| BC047384.1 | - | 3 | 810 | |||