siRNA Details
| Gene Symbol: LOC100996263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121626.1 | 3 | 294 | |||
| GenBank mRNA | BX088863.1 | - | 3 | 235 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440801.5 | - | 3 | 235 | Chr. 1 - 117596832 - 117605711 [-] on Build GRCh38 | |

LOC100996263
Genomic Map