siRNA Details
Gene Symbol: LOC100996263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121626.1 | 3 | 423 | |||
GenBank mRNA | BX088863.1 | - | 3 | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440801.5 | - | 3 | 364 | Chr. 1 - 117596832 - 117605711 [-] on Build GRCh38 |
