siRNA Details
| Gene Symbol: C1orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000062.1 | XP_016855551.1 | 1, 2 | 289 | ||
| XM_017000066.1 | XP_016855555.1 | 1, 2 | 289 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441085.5 | - | 1, 2 | 106 | Chr. 1 - 156446412 - 156456466 [-] on Build GRCh38 | |

C1orf61
Genomic Map