siRNA Details
| Gene Symbol: DSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136520.1 | 2 | 916 | |||
| GenBank mRNA | AK093256.1 | - | 2 | 795 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448740.2 | - | 2 | 861 | Chr. 6 - 116254207 - 116256743 [+] on Build GRCh38 | |

DSE
Genomic Map