siRNA Details
| Gene Symbol: PRMT5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120599.1 | 2 | 1910 | |||
| NR_120600.1 | 2 | 1800 | ||||
| GenBank mRNA | AK093277.1 | - | 2 | 1908 | ||
| DB548852.1 | - | 2 | 252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRMT5-AS1| Gene Symbol: PRMT5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120599.1 | 2 | 1910 | |||
| NR_120600.1 | 2 | 1800 | ||||
| GenBank mRNA | AK093277.1 | - | 2 | 1908 | ||
| DB548852.1 | - | 2 | 252 | |||