siRNA Details
Gene Symbol: PRMT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120599.1 | 1 | 1638 | |||
NR_120600.1 | 1 | 1638 | ||||
GenBank mRNA | AK093277.1 | - | 1 | 1636 | ||
DB548852.1 | - | 1 | 90 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRMT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120599.1 | 1 | 1638 | |||
NR_120600.1 | 1 | 1638 | ||||
GenBank mRNA | AK093277.1 | - | 1 | 1636 | ||
DB548852.1 | - | 1 | 90 |