siRNA Details
| Gene Symbol: LOC105375166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134258.1 | 4 | 2212 | |||
| GenBank mRNA | BX538274.1 | - | 4 | 2212 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451240.1 | - | 4 | 2212 | Chr. 7 - 15667947 - 15681980 [+] on Build GRCh38 | |

LOC105375166
Genomic Map