siRNA Details
Gene Symbol: LOC101927164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110668.1 | 2 | 694 | |||
GenBank mRNA | BC039513.1 | - | 2 | 694 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420350.1 | - | 2 | 694 | Chr. 1 - 224219613 - 224228043 [+] on Build GRCh38 |
