siRNA Details
| Gene Symbol: LOC101927577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110264.1 | 3 | 652 | |||
| NR_110265.1 | 3 | 366 | ||||
| GenBank mRNA | HY009512.1 | - | 3 | 355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927577| Gene Symbol: LOC101927577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110264.1 | 3 | 652 | |||
| NR_110265.1 | 3 | 366 | ||||
| GenBank mRNA | HY009512.1 | - | 3 | 355 | ||