siRNA Details
Gene Symbol: LINC00987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036466.1 | 2 | 854 | |||
GenBank mRNA | AK126248.1 | - | 2 | 462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000427111.4 | - | 2 | 854 | Chr. 12 - 9240003 - 9243039 [+] on Build GRCh38 |
